Key Documents
Z281514
Scienceware® Vikem® pipette aid
pkg of 6 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
vinyl bulb
packaging
pkg of 6 ea
manufacturer/tradename
Bel-Art F37886-0000
volume
85 mL
General description
By squeezing the 85 mL vinyl bulb and releasing, fluid is drawn up into the pipette. The pipette aid is then removed, and the pipette top is quickly covered with a finger to control and release fluid. The tapered PE chuck allows for quick removal when pipette is filled.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)