Key Documents
Z196711
Sigma-Aldrich
Eagle acid/corrosive/flammable-storage cabinets
sliding-door, 45 gal, two shelves
Synonym(s):
chemical containers, chemical storage, flammable cabinets, flammable liquid storage
About This Item
UNSPSC Code:
41100000
NACRES:
JA.01
Pricing and availability is not currently available.
manufacturer/tradename
Eagle
availability
not available in EU
H × W × D
65 in. × 43 in. × 18 in.
General description
Cabinets are FM-approved. All interior and exterior surfaces, including shelves, are finished with blue epoxy powder coating, giving superior protection against chemical attack. Shelf trays and bottom trays of HDPE and each adjustable, 40 W × 143/4 in. D shelf is supported by four brackets and can support more than 500lb. Also includes four leveling legs. Legend: Caution CORROSIVE, in white.
Construction specifications:
All Eagle corrosive and flammable-storage cabinets comply with OSHA 29 CFR 1910. 106d and NFPA Code 30 requirements. Constructed of 18 gauge cold-rolled steel sides, top and bottom; double-walled with 11/2 in. air space between walls and doors. Vents are equipped with flame arresters and removable 2 in. NPT steel plugs; grounding connector is built-in. Equipped with 3-point key lock and 2 in., raised, leak-proof door sill and high-gloss enamel door with red warning legend. For all cabinets, unless otherwise specified, subtract 3 in. from each dimension to get inside dimensions.
Door latchs open by an FM-approved fusible link that melts and automatically closes once the fusible link reaches a temperature of 165°F.
Choose from one of the most recent versions:
Certificates of Analysis (COA)