Skip to Content
Merck
All Photos(2)

Key Documents

W266720

Sigma-Aldrich

L-Menthone

mixture of isomers, natural, ≥96%, FG

Synonym(s):

(-)-Menthone, (1R,4S)-p-Menthan-3-one, (2S,5R)-2-Isopropyl-5-methylcyclohexanone, trans-Menthone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2667
MDL number:
UNSPSC Code:
12352205
Flavis number:
07.176
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
(Peppermint)
food allergen:
no known allergens

biological source

(Peppermint)

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥96%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody; minty

SMILES string

O=C1[C@@H](CC[C@H](C1)C)C(C)C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m1/s1

InChI key

NFLGAXVYCFJBMK-BDAKNGLRSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service