Key Documents
TEM-LFH2100AR1.3
Sigma-Aldrich
QUANTIFOIL on London Finder Grids (H2)
Gold Grids, size 200 mesh, pack of 100
Synonym(s):
Electron Microscopy Grids from QUANTIFOIL
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
size:
200 mesh
packaging:
pack of 100
packaging
pack of 100
size
200 mesh
General description
QUANTIFOIL Gold Grids, 200 Mesh, 1.2 micron hole, 2.5 micron period. London Finder Grids (H2), Pitch 127 micron, 200 mesh, 100/pk
Choose from one of the most recent versions:
Certificates of Analysis (COA)