Key Documents
SYNC204300
Synthware Liebig condenser with 8 mm OD hose connections
joint: ST/NS 24/40, jacket L 300 mm
Synonym(s):
Synthware C204250, Straight condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C204300
jacket L
300 mm
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)