Key Documents
SYNA122440
Synthware vacuum transfer adapter with PTFE stopcocks
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A122440
Related Categories
General description
With 2 mm PTFE stopcock. For transfer of sensitive and hazardous solutions.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)