Skip to Content
Merck
All Photos(2)

Key Documents

P22354

Sigma-Aldrich

trans-2-Phenylcyclopropane-1-carboxylic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C3H4CO2H
CAS Number:
Molecular Weight:
162.19
Beilstein:
3197783
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

86-88 °C (lit.)

SMILES string

OC(=O)[C@@H]1C[C@H]1c2ccccc2

InChI

1S/C10H10O2/c11-10(12)9-6-8(9)7-4-2-1-3-5-7/h1-5,8-9H,6H2,(H,11,12)/t8-,9+/m0/s1

InChI key

AHDDRJBFJBDEPW-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Gerkin
Acta crystallographica. Section C, Crystal structure communications, 53 ( Pt 12), 1989-1991 (1998-02-12)
The title compound, (+/-)-cis-2-phenylcyclopropanecarboxylic acid, C10H10O2, crystallized in the centrosymmetric space group P2(1)/n. The hydrogen bonding is of the cyclic dimer type about a center of symmetry; the Odonor...Oacceptor distance is 2.645 (2) A. The carboxylic H atom is ordered
Edmund J Keliher et al.
Organic & biomolecular chemistry, 4(14), 2777-2784 (2006-07-11)
Many carbenoid cyclopropanation reactions promoted by chiral catalysts give product mixtures reflecting impressive diastereo- and enantioselectivities. Few provide a single chiral product efficiently. This limitation has been overcome in cyclopropanations of styrene and isotopically labeled styrenes with alpha-diazoacetates. Convenient syntheses
R E Gerkin
Acta crystallographica. Section C, Crystal structure communications, 53 ( Pt 9), 1280-1282 (1997-10-06)
The title compound, C10H10O2, crystallized in space group P2(1) with two molecules in the asymmetric unit. In this structure, hydrogen bonding of the cyclic dimer type [with O ... O 2.623(2)-2.637(2)A] links the two molecules of the asymmetric unit. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service