GF15333375
Nickel silver (18) alloy, Cu65Ni18Zn17
100mm tube, 6.35mm outside diameter, 0.5mm wall thickness, 5.35mm inside diameter, hard
Synonym(s):
CZ057750
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 153-333-75
L × thickness
100 mm × 0.5 mm
O.D. × I.D.
6.35 mm × 5.35 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service