DWKCPB0025P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations
class B, capacity 25 mL , with +/-0.5mL tolerance
Synonym(s):
CPB0025P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121805
NACRES:
NA.54
feature
class B
graduated
packaging
pkg of 10 ea
manufacturer/tradename
Azlon CPB0025P
capacity
25 mL , with +/-0.5mL tolerance
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for the measurement and dispensing of liquids in the laboratory
Features and Benefits
Conform to ISO 6706 with anti-suction designto prevent base sticking to wet surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service