DWK45066C-18
KIMBLE® Solid Cap
black phenolic cap (with PTFE liner), cap for 18-415
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Recommended Products
material
PTFE/rubber liner
black phenolic cap (with PTFE liner)
sterility
non-sterile
feature
class B
packaging
case of 225 ea
manufacturer/tradename
Kimble® 45066C-18
fitting
cap for 18-415
General description
Class B Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
- Specially formulated phenolic cap material
- Autoclavable
- White rubber liners
- Excellent for general laboratory use
Features and Benefits
- Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Excellent for general laboratory use
- Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)