Skip to Content
Merck
All Photos(1)

Key Documents

BR781681

BRAND® microplate BRANDplates®, pureGrade

size 384 wells, size 100 μL, flat bottom, white

Synonym(s):

BRANDplates®, Microplates, pureGradeS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
flat bottom
polystyrene
white wells
size:
100 μL
384 wells

material

flat bottom
polystyrene
white wells

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packaged)

manufacturer/tradename

BRAND 781681

size

100 μL
384 wells

working volume

100 μL

color

white

suitability

suitable for (bacteriological assays)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a non-treated surface, making it particularly well-suited for bacteriological assays. Sterile according to ISO 11137 standards, this microplate ensures a contamination-free environment for sensitive experiments. It is classified as BIO-CERT® CELL CULTURE QUALITY, guaranteeing that it is free from human DNA, RNase, DNase, and pyrogens, and is non-cytotoxic. This reliable microplate is an essential tool for researchers conducting microbiological studies, providing consistent performance and safety in cell culture applications.

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service