BR781362
BRAND® PCR Rack Box
Synonym(s):
Accessories, PCR, Rack
About This Item
Recommended Products
material
assorted colors
polypropylene
feature
Holds 96 x 0.2 mL tubes (single, 8-strips or 12-strips)
packaging
pack of 5 ea
manufacturer/tradename
BRAND 781362
technique(s)
PCR: suitable
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service