Key Documents
BR768505
BRAND® tidy tray with compartments
5 compartments
Synonym(s):
Trays
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Pricing and availability is not currently available.
material
PVC
packaging
pack of 1 ea
manufacturer/tradename
BRAND 768505
size
400 mm × 300 mm × 65 mm
General description
The BRAND® tidy tray with compartments is made from durable PVC and is specifically designed for use as a utility tray or drawer organizer. Its compartmentalized structure allows for efficient storage and organization of various laboratory items, such as magnetic stirring bars and other small tools.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)