Key Documents
BAH942404002
Scienceware® Lab Companion 3-way valve
fits all cabinet vacuum desiccators, pack of 1 ea
About This Item
UNSPSC Code:
41104510
Pricing and availability is not currently available.
feature
connects to 1/4 in. tubing
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H94240-4002
General description
Bel-Art Viton 3-Way Valve, available as a replacement of or addition to Lab Companion cabinet-style vacuum desiccators.
Features and Benefits
  • Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange
  • Four built-in ports are standard on the cabinet style units and one Viton® 3-way valve is included; attachment of up to three additional valves is possible
Legal Information
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)