Merck
All Photos(4)
Key Documents
BAF171300024
SP Bel-Art® Pipette Jar for Cleanware Pipette Rinsing System
diam. × H 15.2 cm (6 in.) × 61 cm (24 in.)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F17130-0024
diam. × H
15.2 cm (6 in.) × 61 cm (24 in.)
General description
Store and soak pipettes in these leakproof, molded polyethylene Pipette Jars.
Features and Benefits
  • Withstands all common cleaning solutions at room temperature, including sulfuric acid/dichromate
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP