Key Documents
BAF131930102
Clavies® cover for biohazard bag holder
bench-top
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Pricing and availability is not currently available.
material
polypropylene
feature
autoclavable
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13193-0102
size
203x241 mm
General description
Dishwasher and autoclave safe, the reusable Clavies® Biohazard Bag Holder Cover opens quickly and easily by pressing the built-in release tab – no batteries required! The cover fits both the Medium and Large sizes of the Clavies Biohazard Bag Holders.
  • Opens with a single touch; sanitary, reusable
  • Can be installed in seconds
  • Improves the sanitation of these biohazardous waste containers
  • Center panel of the lid opens to an upright position and reveals a large 20.3 x 24.1cm (8 x 9 1/2 in.) opening through which wastes can be deposited
Legal Information
Clavies is a registered trademark of SP Scienceware, a division of SP
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)