ALD01804
3-((4-chlorophenyl)amino)-3-methylbutan-1-ol
About This Item
form
liquid
storage temp.
2-8°C
SMILES string
CC(CCO)(C)NC1=CC=C(Cl)C=C1
Application
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service