Skip to Content
Merck
All Photos(2)

Key Documents

936111

Sigma-Aldrich

SerrKap Palladacycle

greener alternative

≥95%

Synonym(s):

[Pd(μ-AcO)(4-Cl-CˆN-SO3Na)]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H22Cl2N2Na2O10Pd2S2
CAS Number:
Molecular Weight:
964.36
UNSPSC Code:
12352101
NACRES:
NA.21

Quality Level

Assay

≥95%

form

solid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

yellow

greener alternative category

storage temp.

−20°C

SMILES string

[H]C(C1=C([Pd]23oc(C)o[Pd]4(C5=C(C([H])=[N]4C6=CC=C(S(=O)(O[Na])=O)C=C6)C=CC(Cl)=C5)oc(C)o2)C=C(Cl)C=C1)=[N]3C7=CC=C(S(=O)(O[Na])=O)C=C7

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

SerrKap Palladacycle is a phosphine-free, water soluble Pd dimer catalyst used extensively for low temperature modifications of nucleosides via Suzuki-Miyaura and Heck reactions. It is also compatible with flow reaction conditions and suitable for late-stage oligonucleotide and DNA modification.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh Kori et al.
Current protocols, 2(7), e502-e502 (2022-07-28)
Modification of nucleosides via cross-coupling processes has been carried out extensively on unprotected halonucleosides to produce functionalized nucleosides that are often developed for incorporation into oligonucleotides or used as fluorescent probes. This approach requires protection of the 5'-OH with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service