Skip to Content
Merck
All Photos(1)

Key Documents

929840

Sigma-Aldrich

Poly(ethylene glycol) dimethacrylate

average Mn 3400, contains MEHQ as inhibitor

Synonym(s):

PEG bismethacrylate, PEG hydrogel, PEGDM, PEG dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25
Pricing and availability is not currently available.

form

fine powder

Quality Level

mol wt

average Mn 3400

contains

MEHQ as inhibitor

color

white to off-white

bp

>200 °C/2 mmHg (lit.)

solubility

deionized water: 20 wt. % at 23 °C

Ω-end

methacrylate

α-end

methacrylate

storage temp.

−20°C

SMILES string

OCCO.CC(=C)C(O)=O

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) dimethacrylate is a homobifunctional, hydrophilic, biocompatible polymer commonly used as a crosslinker in PEG hydrogels. Crosslinked networks can be formed via radical initiator induced polymerization or photopolymerization.

Application

PEG dimethacrylates have numerous applications including tissue engineering, 3D printing applications, surface modifications, PEGylation of proteins and peptides, and drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service