Skip to Content
Merck
All Photos(1)

Key Documents

918083

Sigma-Aldrich

2,9-Dimethyl-1,10-phenanthroline nickel (ll) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12Br2N2Ni
CAS Number:
Molecular Weight:
426.76
UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

2,9-Dimethyl-1,10-phenanthroline nickel (ll) bromide is a Ni precatalyst used for a variety of cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing-Yuan Meng et al.
Angewandte Chemie (International ed. in English), 56(43), 13426-13430 (2017-08-24)
We report the efficient carboxylation of bromides and triflates with K2 CO3 as the source of CO2 in the presence of an organic photocatalyst in combination with a nickel complex under visible light irradiation at room temperature. The reaction is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service