Skip to Content
Merck
All Photos(1)

Key Documents

901136

Sigma-Aldrich

1,17-Diazido-3,6,9,12,15-pentaoxaheptadecane

Synonym(s):

1,5-bis[(5-azidopentyl)oxy]pentane, 1-Azido-2-[2-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethane, 3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl bis-azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N6O5
CAS Number:
Molecular Weight:
332.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.4716

density

1.13194 g/mL

storage temp.

2-8°C

SMILES string

[N-]=[N+]=NCCOCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C12H24N6O5/c13-17-15-1-3-19-5-7-21-9-11-23-12-10-22-8-6-20-4-2-16-18-14/h1-12H2

InChI key

OQHIMLOZSDFRID-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service