Skip to Content
Merck
All Photos(1)

Key Documents

797758

Sigma-Aldrich

Gold nano-urchins

100 nm avg. part. size, in 0.1 mM PBS

Synonym(s):

Gold nanoparticles, Non-functionalized gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

description

Particles:∼3.84E+9/mL

Quality Level

form

liquid
nanourchin

OD

1

avg. part. size

100 nm

λmax

680 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Gold nanoparticles are extensively used in biomedical research due to their biocompatibility, facile conjugation to biomolecules and unique optical properties arising from surface plasmon resonance. The presence of spiked nanoantenna-type shapes in Nano-Urchins enhances the plasmonic extinction resonance effect, which improves the optical limiting effects.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiansong Wang et al.
Nanoscale, 4(24), 7766-7772 (2012-11-10)
The hierarchically assembled Au microspheres/sea urchin-like structures have been synthesized in aqueous solution at room temperature with and without proteins (bovine serum albumin, BSA) as mediators. The average diameter of an individual Au microsphere is 300-600 nm, which is composed
Eliza Hutter et al.
ACS nano, 4(5), 2595-2606 (2010-03-25)
Given the emergence of nanotherapeutics and nanodiagnostics as key tools in today's medicine, it has become of critical importance to define precisely the interactions of nanomaterials with biological systems and to characterize the resulting cellular response. We report here the
Eliza Hutter et al.
Microscopy research and technique, 74(7), 592-604 (2010-09-11)
Nanoparticles are the latest tool acquired by the science of bioimaging, serving primarily as new contrast agents, sensors, or signal enhancing agents in established and developing imaging techniques. This review focuses on the unique properties of two classes of nanoparticles:

Articles

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service