Skip to Content
Merck
All Photos(2)

Key Documents

777056

Sigma-Aldrich

5′-Bromo-2,2′-bithiophene-5-carboxaldehyde

97%

Synonym(s):

5′-Bromo-2,2′-bithienyl-5-carboxaldehyde, 5′-Bromo-2,2′-bithiophene-5-carbaldehyde, 5′-Bromo-[2,2′]bithiophene-5-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5BrOS2
CAS Number:
Molecular Weight:
273.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

140-145 °C

functional group

aldehyde
bromo

storage temp.

2-8°C

SMILES string

BrC1=CC=C(C2=CC=C(C([H])=O)S2)S1

InChI

1S/C9H5BrOS2/c10-9-4-3-8(13-9)7-2-1-6(5-11)12-7/h1-5H

InChI key

NTHMTYNJFSUBMF-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service