Skip to Content
Merck
All Photos(1)

Key Documents

724580

Sigma-Aldrich

1-(4-bromophenyl)piperazine

95%

Synonym(s):

1-(4-bromophenyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13BrN2
CAS Number:
Molecular Weight:
241.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

solid

mp

91-95 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

Brc1ccc(cc1)N2CCNCC2

InChI

1S/C10H13BrN2/c11-9-1-3-10(4-2-9)13-7-5-12-6-8-13/h1-4,12H,5-8H2

InChI key

PJHPFAFEJNBIDC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Liu et al.
Journal of pharmaceutical and biomedical analysis, 117, 325-332 (2015-09-29)
The current study reports the development and validation of a stability-indicating reversed phase HPLC method for the separation and identification of potential impurities in vortioxetine, a recently developed antidepressant. The structures of a new compound and four process-related impurities formed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service