Skip to Content
Merck
All Photos(1)

Documents

698253

Sigma-Aldrich

Tris(3,5-dimethylphenyl)phosphine

96%

Synonym(s):

Tri-3,5-xylylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2C6H3]3P
CAS Number:
Molecular Weight:
346.44
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

160-165 °C

functional group

phosphine

SMILES string

Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3cc(C)cc(C)c3

InChI

1S/C24H27P/c1-16-7-17(2)11-22(10-16)25(23-12-18(3)8-19(4)13-23)24-14-20(5)9-21(6)15-24/h7-15H,1-6H3

InChI key

XRALRSQLQXKXKP-UHFFFAOYSA-N

Application

Catalyst for:
  • Cu / diphosphine-catalyzed asymmetric hydrogenation of heteroaromatic ketones and enones
  • Highly selective rhodium-catalyzed hydrogenation reactions
  • Classical versus kinetic resolution in preparation of privileged silicon-stereogenic silanaphthalenes

Used for kinetic resolution of donor-functionalized secondary alcohols via Cu-H-catalyzed stereoselective silylation by dehydrogenative Si-O coupling with Si-stereogenic silanes

Used for comparative studies of conformational rigidity of silicon-stereogenic silanes in asymmetric catalysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daichi Kakizoe et al.
Dalton transactions (Cambridge, England : 2003), 46(43), 14804-14811 (2017-10-19)
We report the synthesis and photophysical properties of Cu(i) complexes with 1,10-phenanthroline (phen) and monodentate phosphine ligands. Single crystal X-ray structural analysis revealed that these have three-coordinated trigonal planar geometries. We also found that one of them, [Cu(phen)(Johnphos)]BF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service