Skip to Content
Merck
All Photos(1)

Documents

668338

Sigma-Aldrich

9-Undecyn-1-ol

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O
CAS Number:
Molecular Weight:
168.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.466

bp

92-93 °C/0.03 mmHg

density

0.866 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC#CCCCCCCCCO

InChI

1S/C11H20O/c1-2-3-4-5-6-7-8-9-10-11-12/h12H,4-11H2,1H3

InChI key

YSDWKSIWEWYTEF-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service