Skip to Content
Merck
All Photos(1)

Key Documents

641367

Sigma-Aldrich

Bis(4-methoxyphenyl) disulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OC6H4S)2
CAS Number:
Molecular Weight:
278.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

41-45 °C (lit.)

functional group

disulfide

SMILES string

COc1ccc(SSc2ccc(OC)cc2)cc1

InChI

1S/C14H14O2S2/c1-15-11-3-7-13(8-4-11)17-18-14-9-5-12(16-2)6-10-14/h3-10H,1-2H3

InChI key

PZQGLCGLPMWYBT-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service