Skip to Content
Merck
All Photos(1)

Key Documents

588652

Sigma-Aldrich

4-Hydroxybenzaldehyde-d6

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DO-C6D4-CDO
CAS Number:
Molecular Weight:
128.16
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

isotopic purity

99 atom % D

form

solid

mp

117-119 °C (lit.)

mass shift

M+6

SMILES string

[2H]Oc1c([2H])c([2H])c(C([2H])=O)c([2H])c1[2H]

InChI

1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H/i1D,2D,3D,4D,5D/hD

InChI key

RGHHSNMVTDWUBI-QNKSCLMFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service