Skip to Content
Merck
All Photos(3)

Key Documents

552003

Sigma-Aldrich

D-Glucose-1,2,3,4,5,6,6-d7

97 atom % D

Synonym(s):

Deuterated Glucose, Labeled Glucose, D-Glucose-C-d7, Dextrose-C-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5D7O6
CAS Number:
Molecular Weight:
187.20
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

97 atom % D

Quality Level

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

technique(s)

bio NMR: suitable
protein expression: suitable

mp

150-152 °C (dec) (lit.)

mass shift

M+7

storage temp.

room temp

SMILES string

[2H]C([2H])(O)[C@@]1([2H])OC([2H])(O)[C@]([2H])(O)[C@@]([2H])(O)[C@]1([2H])O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4-,5+,6?/m0/s1/i1D2,2D,3D,4D,5D,6D

InChI key

WQZGKKKJIJFFOK-KMTOWNPSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

D-Glucose-1,2,3,4,5,6,6-d7 is a stable isotopes and simple sugar containing six carbon atoms in the cyclic form.

Application

D-glucose-1,2,3,4,5,6,6-d7 can be used:
  • As a dopant in the process of controlling the quality of grape juice contaminated by apple juice using electrospray ionization Fourier transform ion cyclotron resonance mass spectrometry [ESI (-) FT-ICR MS].
  • To quantify the physical and chemical recycling of glucose by measuring the difference in their metabolism.
  • In combination with D2O to increase the lipid production by reducing the D-H exchange.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conor C Horgan et al.
Journal of materials chemistry. B, 8(20), 4447-4459 (2020-05-07)
Extracellular vesicles (EVs) are biologically-derived nanovectors important for intercellular communication and trafficking. As such, EVs show great promise as disease biomarkers and therapeutic drug delivery vehicles. However, despite the rapidly growing interest in EVs, understanding of the biological mechanisms that
Qingci Zhao et al.
Cell reports, 32(8), 108074-108074 (2020-08-28)
The small guanosine triphosphatase (GTPase) RAS serves as a molecular switch in signal transduction, and its mutation and aberrant activation are implicated in tumorigenesis. Here, we perform real-time, in-cell nuclear magnetic resonance (NMR) analyses of non-farnesylated RAS to measure time
Jue Hou et al.
Biophysical journal, 119(2), 258-264 (2020-07-02)
Investigating the behavior of breast cancer cells via reaction kinetics may help unravel the mechanisms that underlie metabolic changes in tumors. However, obtaining human in vivo kinetic data is challenging because of difficulties associated with measuring these parameters. Nondestructive methods of

Articles

Utilizing ISOGRO® Supplementation of M9 Minimal Media to Enhance Recombinant Protein Expression.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service