All Photos(1)
About This Item
Linear Formula:
F3CSC6H4CHO
CAS Number:
Molecular Weight:
206.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
90%
impurities
~1% 4-(Trifluoromethylthio)benzoic acid
~5% 4-(Trifluoromethylthio)benzonitrile
refractive index
n20/D 1.511 (lit.)
bp
219 °C (lit.)
mp
29-31 °C (lit.)
functional group
aldehyde
fluoro
thioether
SMILES string
FC(F)(F)Sc1ccc(C=O)cc1
InChI
1S/C8H5F3OS/c9-8(10,11)13-7-3-1-6(5-12)2-4-7/h1-5H
InChI key
OPAXHVNZGUGCKW-UHFFFAOYSA-N
Related Categories
General description
Enthalpy of vaporization of 4-(trifluoromethylthio)benzaldehyde at boiling point (492.15K) is 43.130kjoule/mol.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yaws CL
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service