Skip to Content
Merck
All Photos(3)

Key Documents

489735

Sigma-Aldrich

Hydrazine sulfate-15N2

98 atom % 15N

Synonym(s):

15N Labeled hydrazine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215N15NH2 · H2SO4
CAS Number:
Molecular Weight:
132.11
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % 15N

Quality Level

form

solid

mp

254 °C (lit.)

density

1.391 g/mL at 25 °C

mass shift

M+2

storage temp.

room temp

SMILES string

[15NH2][15NH2].OS(O)(=O)=O

InChI

1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)/i1+1,2+1;

InChI key

ZGCHATBSUIJLRL-AWQJXPNKSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service