Skip to Content
Merck
All Photos(2)

Key Documents

446521

Sigma-Aldrich

Poly[N,N′-(1,3-phenylene)isophthalamide]

Synonym(s):

1,3-Benzenediamine-1,3-benzenedicarbonyl dichloride copolymer, 1,3-Benzenediamine-isophthalyl chloride copolymer, m -Diaminobenzene-isophthaloyl chloride copolymer, m -Phenylenediamine-isophthaloyl chloride copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C14H14N2O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

mp

>371 °C (dec.) (lit.)

solubility

DMF-LiCl: soluble

density

1.38 g/mL at 25 °C (lit.)

SMILES string

c1ccccc1.NC(=O)c2cccc(c2)C(N)=O

InChI

1S/C8H8N2O2.C6H6/c9-7(11)5-2-1-3-6(4-5)8(10)12;1-2-4-6-5-3-1/h1-4H,(H2,9,11)(H2,10,12);1-6H

InChI key

FTTVLDNWOCMNMX-UHFFFAOYSA-N

Application

Used in heat and flame protective clothing. Electrical insulating paper requiring high temperature stability.

Physical properties

Does not melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service