Skip to Content
Merck
All Photos(2)

Key Documents

369047

Sigma-Aldrich

(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide

98%

Synonym(s):

NSC 82468

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CCH2P(C6H5)3Br
CAS Number:
Molecular Weight:
459.36
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

178 °C (dec.) (lit.)

functional group

ester
phosphine

SMILES string

[Br-].CC(C)(C)OC(=O)C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C24H26O2P.BrH/c1-24(2,3)26-23(25)19-27(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22;/h4-18H,19H2,1-3H3;1H/q+1;/p-1

InChI key

ZGLFRTJDWWKIAK-UHFFFAOYSA-M

Application

Reactant for:
  • Rhodium-catalyzed asymmetric hydrogenation reactions
  • Wittig reaction
  • Wittig methylenation
  • Preparation of N-(phenylmethyl)-cis-pyrrolidinediacetic acid esters via double aza-Michael addition reaction
  • Stereoselective preparation of of α-fluoro-α,β-unsaturated esters via deprotonation, followed by fluorination and stereoselective Wittig olefination with aldehydes
  • Wittig chain extension reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service