Skip to Content
Merck
All Photos(1)

Key Documents

346489

Sigma-Aldrich

3-(Trifluoromethoxy)benzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CHO
CAS Number:
Molecular Weight:
190.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

83-86 °C/24 mmHg (lit.)

density

1.33 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

FC(F)(F)Oc1cccc(C=O)c1

InChI

1S/C8H5F3O2/c9-8(10,11)13-7-3-1-2-6(4-7)5-12/h1-5H

InChI key

FQEVHRCPXFKJHF-UHFFFAOYSA-N

Application

3-(Trifluoromethoxy)benzaldehyde may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service