Skip to Content
Merck
All Photos(1)

Documents

287784

Sigma-Aldrich

(1R,2R,3S,5R)-(−)-Pinanediol

99%

Synonym(s):

(−)-2-Hydroxyisopinocampheol, [1R-(1α,2α,3α,5α)]-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Beilstein:
2039144
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]21/D −8.6°, c = 6.5 in toluene

optical purity

ee: 97% (GLC)

bp

101-102 °C/1 mmHg (lit.)

mp

57-59 °C (lit.)

SMILES string

CC1(C)[C@H]2C[C@H](O)[C@](C)(O)[C@@H]1C2

InChI

1S/C10H18O2/c1-9(2)6-4-7(9)10(3,12)8(11)5-6/h6-8,11-12H,4-5H2,1-3H3/t6-,7-,8+,10-/m1/s1

InChI key

MOILFCKRQFQVFS-BDNRQGISSA-N

Application

Used to prepare chiral double allylation reagents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple, stable, and versatile double-allylation reagents for the stereoselective preparation of skeletally diverse compounds.
Feng Peng et al.
Journal of the American Chemical Society, 129(11), 3070-3071 (2007-02-24)
Gerald I Richard et al.
Applied spectroscopy, 62(5), 476-480 (2008-05-24)
The spectroscopic properties of a chiral boronic acid based resorcinarene macrocycle employed for chiral analysis were investigated. Specifically, the emission and excitation characteristics of tetraarylboronate resorcinarene macrocycle (TBRM) and its quantum yield were evaluated. The chiral selector TBRM was investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service