Skip to Content
Merck
All Photos(2)

Documents

182869

Sigma-Aldrich

Poly(styrene-co-acrylonitrile)

average Mw ~185,000 by GPC, acrylonitrile 30 wt. %, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH(CN)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mw ~185,000 by GPC

composition

acrylonitrile, 30 wt. %

density

1.08 g/mL at 25 °C (lit.)

SMILES string

C=CC#N.C=Cc1ccccc1

InChI

1S/C8H8.C3H3N/c1-2-8-6-4-3-5-7-8;1-2-3-4/h2-7H,1H2;2H,1H2

InChI key

SCUZVMOVTVSBLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Acrylonitrile-styrene-acrylate particles with different microstructure for improving the toughness of poly (styrene-co-acrylonitrile) resin: Examines the enhancement of SAN resin toughness using ASA particles with various morphologies, potentially useful for developing advanced polymeric materials (Y Zhang et al., 2021).
  • Fabrication by spin-coating and optical characterization of poly (styrene-co-acrylonitrile) thin films: Describes the fabrication and optical properties assessment of PSAN thin films, useful in materials science for applications requiring detailed optical properties analysis (E Hedl et al., 2021).
  • Rapid fabrication of periodic patterns on poly (styrene-co-acrylonitrile) surfaces using direct laser interference patterning: Discusses a method for patterning PSAN surfaces, relevant for materials science research involving microfabrication and nanotechnology (MF Broglia et al., 2015).
  • A novel solid-solid phase change material based on poly (styrene-co-acrylonitrile) grafting with palmitic acid copolymers: Investigates PSAN-based solid-solid phase change materials, which could be significant in drug delivery systems where temperature control is crucial (S Mu et al., 2015).
  • Synthesis, optimization, and characterization of poly (Styrene‐co‐Acrylonitrile) copolymer prepared via precipitation polymerization: Provides insights into the synthesis and properties of PSAN copolymers, applicable in designing materials with specific mechanical and thermal characteristics (MR El‐Aassar et al., 2018).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hygienic chemical studies on plastics (II). Residual acrylonitrile in ABS resin or AS resin and its migration into water (author's transl)].
T Tatsuno et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (97)(97), 93-97 (1979-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service