Z363499
Pipette workstation
stainless steel
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art H18962-0006
Description générale
All-steel universal 5-position rack holds most popular brands and sizes of pipettes.