Merck
P7613
Scienceware® plate reader
mirror magnification 3X actual plate size
Synonyme(s) :
378610000, F37861-0000, cell counting device, cell reader, hands freee cell reader, magnified cell reader
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.13
UNSPSC Code:
41161502
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37861-0000
color
none
General description
The plate reader has a clear deck for resting plates and a large, clear, adjustable mirror for viewing their underside. This tool allows safe examination of the bottom of the plates without risking spills on the face and eyes. It is ideal for Hemagglutination, Hemagglutination Inhibition, Hemolysis and Agglutination testing. Other applications include counting of bacterial cell cultures for colony or plaque growth. Mirror magnification is 3X actual plate size. Polypropylene with an acrylic deck.
Features and Benefits
  • Easy, magnified viewing; hands-free
  • For microplates, view color changes and locate cells for further analysis
  • For petri dishes, view cell growth and take advantage of hands-free design to make notes and isolate cell colonies
  • Ideal for evaluating monolayer cell growth
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP