Merck
HS81121D
Inoculating Needle/Loops
loop volume 10 μL, Rigid, sterile, blue
Synonyme(s) :
Inoculating loops, Inoculation needles
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.51
UNSPSC Code:
41121800
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
acrylonitrile-butadiene-styrene (ABS) , blue
sterility
sterile
feature
Rigid
packaging
pack of 1000 ea
manufacturer/tradename
Heathrow Scientific HS81121D
loop volume
10 μL
General description
The Inoculating Needle/Loops combination provides choice to spread or pick colonies and cultures. Transfer bacterial cultures without risk of contamination.
Features and Benefits
  • Use loop end to uniformly streak a gel surface
  • Use needle end for removal of colonies
  • Constructed of rigid ABS
  • Sterile, 20 per bag with lot-to-lot traceability
  • Disposable
Still not finding the right product?
Explore all of our products under Inoculating Needle/Loops