Merck
61341
D-Lactose monohydrate
tested according to Ph. Eur.
Synonyme(s) :
β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Sucre de lait
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
Formule empirique (notation de Hill) :
C12H22O11 · H2O
Numéro CAS:
Poids moléculaire :
360.31
EC Number:
200-559-2
UNSPSC Code:
12352201
PubChem Substance ID:
Beilstein/REAXYS Number:
3768231
MDL number:
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Quality Segment
agency
tested according to Ph. Eur.
form
solid
storage temp.
room temp
SMILES string
O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O
InChI
1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1
InChI key
HBDJFVFTHLOSDW-XBLONOLSSA-N
Biochem/physiol Actions
Lactose is a dissacharide formed by the condensation of one galactose and one glucose molecule. Lactose is the major sugar in the milk of most species, typically present between 2-8%. The enzyme lactase hydrolyzes lactose to its constituent monosaccharides. In neonates, glucose released via the action of lactase is a major energy source.
Other Notes
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Still not finding the right product?
Explore all of our products under D-Lactose monohydrate
Classe de stockage
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe