Merck
  • A review of loratadine in the treatment of patients with allergic bronchial asthma.
A review of loratadine in the treatment of patients with allergic bronchial asthma.
Clinical therapeutics (1998-01-28)
J L Menardo, F Horak, M R Danzig, W Czarlewski
ABSTRACT
Patients with seasonal or allergic bronchial asthma experience an immediate allergic response caused by allergen-specific immunoglobulin E-mediated histamine release. The release of histamine and other chemical mediators may trigger airway hyperresponsiveness and exaggerated bronchoconstriction, characteristic features of allergic bronchial asthma. Traditional antihistamines have demonstrated only moderate activity of short duration against this disease. In contrast, loratadine, a potent, nonsedating, histamine-1-receptor antagonist with activity in seasonal and perennial allergic rhinitis, has demonstrated effective control of asthma symptoms, improved pulmonary function, and long duration of action in patients with allergic bronchial asthma. This review summarizes preclinical evidence for the antiallergic activity of loratadine and the results of clinical studies with oral loratadine in patients with allergic bronchial asthma.
Sigma-Aldrich
Loratadine, ≥98% (HPLC), powder
Supelco
Loratadine, Pharmaceutical Secondary Standard; Certified Reference Material
Loratadine for system suitability, European Pharmacopoeia (EP) Reference Standard
Loratadine, European Pharmacopoeia (EP) Reference Standard
USP
Loratadine, United States Pharmacopeia (USP) Reference Standard