Merck
  • Multiresidue method for determination of 88 pesticides in berry fruits using solid-phase extraction and gas chromatography-mass spectrometry: Determination of 88 pesticides in berries using SPE and GC-MS.
Multiresidue method for determination of 88 pesticides in berry fruits using solid-phase extraction and gas chromatography-mass spectrometry: Determination of 88 pesticides in berries using SPE and GC-MS.
Food chemistry (2011-07-15)
X Yang, H Zhang, Y Liu, J Wang, Y C Zhang, A J Dong, H T Zhao, C H Sun, J Cui
ABSTRACT
A method using solid phase extraction (SPE) cleanup followed by gas chromatography-mass spectrometry (GC-MS) has been established for quantitative determination of 88 pesticide residues in berry fruits including raspberry, strawberry, blueberry and grape. Based on an appraisal of the characteristics of GC-MS, validation experiments were conducted for 88 pesticides. In the method, solid-phase extraction was carried out using Envi-Carb cartridge coupled with NH(2)-LC cartridge with acetonitrile-toluene (3:1, v/v) as the eluted solvent. In the linear range of each pesticide, the correlation coefficient was R(2)⩾0.99. At the low, medium and high three fortification levels of 0.05-0.5mgkg(-1), recoveries fell within 63-137%. The relative standard deviation was between 1% and 19% for all 88 pesticides. Low limits of detection (0.006-0.05mgkg(-1)) and quantification (0.02-0.15mgkg(-1)) were readily achieved with this method for all tested pesticides.
Product Description
Supelco
Cycloate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible