Merck
  • Effect of catechins and high-temperature-processed green tea extract on scavenging reactive oxygen species and preventing Aβ1-42 fibrils' formation in brain microvascular endothelium.
Effect of catechins and high-temperature-processed green tea extract on scavenging reactive oxygen species and preventing Aβ1-42 fibrils' formation in brain microvascular endothelium.
Nutritional neuroscience (2018-08-17)
Seon-Bong Lee, Eun-Hye Choi, Kang-Hyun Jeong, Kwang-Sik Kim, Soon-Mi Shim, Gun-Hee Kim
ABSTRACT
The present study investigated the effect of high-temperature-processed green tea extract (HTP_GTE) and its bioactive components on the reduction of reactive oxygen species (ROS) and amyloid-beta (Aβ) protein in human microvascular endothelial cells. Compared to Aβ1-42-only treatment, pretreatment of HTP_GTE was revealed to effectively inhibit ROS generation (P<0.05). HTP_GTE and catechins not only inhibit Aβ1-42 fibril formation but also destabilize preformed Aβ1-42 fibrils. The presence of HTP_GTE, Aβ1-42 fibril formation was significantly inhibited in a dose-dependent manner at 12.5-100 μg/ml of HTP_GTE, showing 86-56%, respectively. Treatment of various concentrations of HTP_GTE and catechins steadily destabilized the preformed Aβ1-42 fibrils for 24 h in a dose-dependent manner. It was observed that the gallated groups such as epigallocatechin gallate, epicatechin gallate, gallocatechin gallate, and catechin gallate more effectively disturbed Aβ1-42 fibril formation and destabilized the preformed Aβ1-42 fibrils than the non-gallated group. Taken together, these findings supported that sterilized green tea could be promising natural anti-amyloidogenic agents associated with therapeutic approaches in Alzheimer's disease by scavenging ROS generation and Aβ fibril in the brain tissue.
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(−)-Catechin gallate, ≥98% (HPLC), from green tea