
(−)-Catechin gallate

Synonym(s):
(2S,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-(3,4,5-trihydroxybenzoate)
Empirical Formula (Hill Notation):
C22H18O10
CAS Number:
Molecular Weight:
442.37
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Application
Showing 1-1 of 1