Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H11N3O4S
CAS Number:
Molecular Weight:
317.32
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
API family
piroxicam
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecules)
format
neat
InChI
1S/C14H11N3O4S/c18-13-9-5-1-2-6-10(9)22(20,21)17-12(13)14(19)16-11-7-3-4-8-15-11/h1-8,17-18H,(H,15,16,19)
InChI key
XRXYFCIBSPQECM-UHFFFAOYSA-N
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - STOT RE 2 Oral
target_organs
Gastrointestinal tract
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

