Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H28N4O6 · 2HCl
CAS Number:
Molecular Weight:
517.40
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
grade
pharmaceutical primary standard
agency
USP
API family
mitoxantrone
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Cl[H].Cl[H].OCCNCCNc1ccc(NCCNCCO)c2C(=O)c3c(O)ccc(O)c3C(=O)c12
InChI
1S/C22H28N4O6.2ClH/c27-11-9-23-5-7-25-13-1-2-14(26-8-6-24-10-12-28)18-17(13)21(31)19-15(29)3-4-16(30)20(19)22(18)32;;/h1-4,23-30H,5-12H2;2*1H
InChI key
ZAHQPTJLOCWVPG-UHFFFAOYSA-N
Gene Information
human ... TOP2A(7153)
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Mitoxantrone hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Mitoxantrone Injection
Biochem/physiol Actions
Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Mitoxantrone hydrochloride
signalword
Danger
hcodes
Hazard Classifications
Muta. 1B - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
