Skip to Content
Merck

1370101

USP

Lopinavir

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H48N4O5
CAS Number:
Molecular Weight:
628.80
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

grade

pharmaceutical primary standard

agency

USP

API family

lopinavir

form

powder

packaging

pkg of 350 mg

manufacturer/tradename

USP

storage condition

protect from light

color

white to off-white

mp

255.2-260.6 °F (124—127°C)

solubility

chloroform: soluble, isopropanol: soluble, methanol: freely soluble, methylene chloride: freely soluble, water: practically insoluble

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N4(CCCNC4=O)[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](NC(=O)COc3c(cccc3C)C)Cc2ccccc2)Cc1ccccc1

InChI

1S/C37H48N4O5/c1-25(2)34(41-20-12-19-38-37(41)45)36(44)39-30(21-28-15-7-5-8-16-28)23-32(42)31(22-29-17-9-6-10-18-29)40-33(43)24-46-35-26(3)13-11-14-27(35)4/h5-11,13-18,25,30-32,34,42H,12,19-24H2,1-4H3,(H,38,45)(H,39,44)(H,40,43)/t30-,31-,32-,34-/m0/s1

InChI key

KJHKTHWMRKYKJE-SUGCFTRWSA-N

General description

Lopinavir is an antiretroviral agent and a protease inhibitor indicated for the management of human immunodeficiency virus (HIV) infection. It is a key component of Kaletra, the co-formulation of lopinavir and ritonavir.

Application

Lopinavir may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and plasma samples by chromatography techniques.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Lopinavir


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library