Skip to Content
Merck

Skip To

1356971

USP

Letrozole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4,4′-(1H-1,2,4-Triazol-1-ylmethylene)bisbenzonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H11N5
CAS Number:
Molecular Weight:
285.30
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

letrozole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N#CC(C=C1)=CC=C1C(N2C=NC=N2)C3=CC=C(C#N)C=C3

InChI

1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H

InChI key

HPJKCIUCZWXJDR-UHFFFAOYSA-N

Gene Information

human ... CYP19A1(1588)

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Letrozole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Letrozole Tablets

Biochem/physiol Actions

Letrozole is a non-steroidal aromatase inhibitor.
Letrozole is a third generation nonsteroidal aromatase inhibitor. It is a competitive inhibitor of the aromatase enzyme system and thus inhibits the conversion of androgens to estrogens. Letrozole inhibits the aromatase enzyme by competitively binding to the heme of the cytochrome P450 subunit of the enzyme, resulting in a reduction of estrogen biosynthesis in all tissues.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000685BP1234L6545
Letrozole British Pharmacopoeia (BP) Reference Standard

BP1234

Letrozole

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

format

neat

format

neat

format

neat

format

-

Gene Information

human ... CYP19A1(1588)

Gene Information

human ... CYP19A1(1588)

Gene Information

human ... CYP19A1(1588)

Gene Information

human ... CYP19A1(1588)

API family

letrozole

API family

letrozole

API family

letrozole

API family

-


Still not finding the right product?

Explore all of our products under Letrozole


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Repr. 2 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters