Skip to Content
Merck

Skip To

1356891

USP

Lauryl alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Dodecanol, Alcohol C12, Dodecyl alcohol, Lauryl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)11OH
CAS Number:
Molecular Weight:
186.33
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1738860
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

7.4 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

API family

lauryl alcohol

autoignition temp.

500 °F

expl. lim.

4 %

manufacturer/tradename

USP

refractive index

n20/D 1.442 (lit.)

bp

260-262 °C (lit.)

mp

22-26 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCO

InChI

1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

LQZZUXJYWNFBMV-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lauryl alcohol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Stearyl Alcohol
  • Cetyl Alcohol
  • Cetyl Esters Wax
  • Myristyl Alcohol
  • Cetostearyl Alcohol
  • Cocoyl Caprylocaprate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W261718PHR25991356905
grade

pharmaceutical primary standard

grade

FG, Halal, Kosher

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

flavors and fragrances

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

expl. lim.

4 %

expl. lim.

4 %

expl. lim.

-

expl. lim.

-

vapor pressure

0.1 mmHg ( 20 °C)

vapor pressure

0.1 mmHg ( 20 °C)

vapor pressure

-

vapor pressure

-


Still not finding the right product?

Explore all of our products under Lauryl alcohol


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

249.8 °F - closed cup

flash_point_c

121 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters