Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
itraconazole
manufacturer/tradename
USP
application(s)
pharmaceutical
pharmaceutical (small molecule)
format
mixture
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Itraconazole
- Itraconazole Capsules
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description Pharmaceutical Analytical Impurity (PAI) | description United States Pharmacopeia (USP) Reference Standard | description Pharmaceutical Analytical Impurity (PAI) |
| grade pharmaceutical primary standard | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical primary standard | grade pharmaceutical analytical impurity (PAI) |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| format mixture | format - | format mixture | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule |
| API family itraconazole | API family - | API family abiraterone | API family - |
| agency USP | agency USP | agency USP | agency USP |
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
